missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPN1 / XAB1 (DNA Repair & Protein Synthesis) Mouse, Clone: GPN1/2350, enQuireBio™
Specifications
Specifications
| Antigen | GPN1 / XAB1 (DNA Repair & Protein Synthesis) |
| Applications | ELISA, Immunohistochemistry |
| Cellular Localization | Cytoplasmic and Nuclear |
| Classification | Monoclonal |
| Clone | GPN1/2350 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human GPN1 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?