missing translation for 'onlineSavingsMsg'
Learn More

GPN1 / XAB1 (DNA Repair & Protein Synthesis) Mouse, Clone: GPN1/2350, enQuireBio™

Product Code. p-7155986
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15999797 20 μg 20µg
15909807 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15999797 Supplier EnquireBio Supplier No. 11321MSM1P0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen GPN1 / XAB1 (DNA Repair & Protein Synthesis)
Applications ELISA, Immunohistochemistry
Cellular Localization Cytoplasmic and Nuclear
Classification Monoclonal
Clone GPN1/2350
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Host Species Mouse
Immunogen Recombinant full-length human GPN1 protein
Molecular Weight of Antigen 42kDa
Positive Control Pancreas, Testis, Thyroid or Bladder.
Purification Method Protein A/G purified from Bioreactor concentrate.
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11321
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.