Learn More
MGMT Antibody (MGMT/4791), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21103B
Product Code. 18671085
  • € 726.00 / 0.10mL
Description
MGMT Monoclonal antibody specifically detects MGMT in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MGMT
Monoclonal
Biotin
6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase
Purified His-tagged MGMT protein
0.1 mL
Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors
4255
Store at 4°C in the dark.
IgG1
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
MGMT/4791
PBS
Mouse
Protein A or G purified