missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPN3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00051184-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MPRYAQLVMGPAGSGKSTYCATMVQHCEALNRSVQVVNLDPAAEHFNYSVMADIRELIEVDDVMEDDSLRFGPNGGLVFCMEYFANNFDWLENCLGHVEDDYILFDCPGQIELYTHLPVMKQLVQQLEQWEFRVCGVFLVDSQFMVESFKFISGILAALSAMISLEIPQVNIMTKMDLLSKKAKKEIEKFLDPDMYSLLEDSTSDLRSKKFKKLTKAICGLIDDYSMVRFLPYDQSDEESMNIALQHIDFAIQYGEDLEFKEPKEREDESSSMFDEYFQECQDESpecifications
| GPN3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_016301 | |
| GPN3 | |
| ATPBD1C (NP_057385, 1 a.a. ∼ 284 a.a) full-length human protein. | |
| RUO | |
| 51184 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| GPN-loop GTPase 3 | |
| ATPBD1C/MGC14560/MGC32810 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction