Learn More
GPR101 Antibody (909603) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB7495L
Product Code. 18563576
  • € 644.00 / 0.10mL
Description
GPR101 Monoclonal antibody specifically detects GPR101 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR101
Monoclonal
DyLight 550
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
909603
Western Blot, Flow Cytometry, CyTOF-ready
G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101
NS0 mouse myeloma cell line transfected with human GPR101, Met1-Pro508,, Accession # NP_473362
0.1 mL
GPCR
83550
Store at 4°C in the dark.