missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR101 Antibody (909603) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7495F
This item is not returnable.
View return policy
Description
GPR101 Monoclonal antibody specifically detects GPR101 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| GPR101 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 909603 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101 | |
| NS0 mouse myeloma cell line transfected with human GPR101, Met1-Pro508,, Accession # NP_473362 | |
| 0.1 mL | |
| GPCR | |
| 83550 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction