missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR101 Antibody (909603), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7495PECY55
This item is not returnable.
View return policy
Description
GPR101 Monoclonal antibody specifically detects GPR101 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR101 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 909603 | |
| Flow Cytometry | |
| G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101 | |
| NS0 mouse myeloma cell line transfected with human GPR101, Met1-Pro508,, Accession # NP_473362 | |
| 0.1 mL | |
| GPCR | |
| 83550 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction