missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR109B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15304451
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15304451 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15304451 Supplier Invitrogen™ Supplier No. PA533607

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR109B Polyclonal Antibody for IHC (P)

Receptor for 3-OH-octanoid acid mediates a negative feedback regulation of adipocyte lipolysis to counteract prolipolytic influences under conditions of physiological or pathological increases in beta-oxidation rates. Acts as a low affinity receptor for nicotinic acid. This pharmacological effect requires nicotinic acid doses that are much higher than those provided by a normal diet. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR109B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene HCAR3
Gene Accession No. P49019
Gene Alias G protein-coupled receptor 109B; GPR109B; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74B; GTP-binding protein; HCA3; HCAR3; HM74; HM74B; hydroxycarboxylic acid receptor 3; hydroxy-carboxylic acid receptor 3; Niacin receptor 2; NIACR2; Nicotinic acid receptor 2; PUMAG; Puma-g; putative chemokine receptor
Gene Symbols HCAR3
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-terminus of human GPR109B / HM74.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8843
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.