missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR111 Antibody (594522), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB64941APCCY7
This item is not returnable.
View return policy
Description
GPR111 Monoclonal antibody specifically detects GPR111 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR111 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 594522 | |
| Flow Cytometry | |
| G protein-coupled receptor 111 | |
| Chinese Hamster Ovary cell line, CHO-derived human GPR111, Cys19-Lys375, Accession # Q8IZF7 | |
| 0.1 mL | |
| GPCR | |
| 222611 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction