Learn More
GPR111 Antibody (594522), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB64941K
This item is not returnable.
View return policy
Description
GPR111 Monoclonal antibody specifically detects GPR111 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| GPR111 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 594522 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 111 | |
| Chinese Hamster Ovary cell line, CHO-derived human GPR111, Cys19-Lys375, Accession # Q8IZF7 | |
| 0.1 mL | |
| GPCR | |
| 222611 | |
| Store at 4°C in the dark. | |
| IgG2b |