missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR111 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557299
This item is not returnable.
View return policy
Description
Immunogen sequence: CDGVCTDYSQ CTQPCPPDTQ GNMGFSCRQK TWHKITDTCR TLNALNIFEE DSRLVQPFED NIKISVYTGK SETITDMLLQ KCPTD Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 38%.
Specifications
| GPR111 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF2 | |
| ADGRF2; adhesion G protein-coupled receptor F2; adhesion G-protein coupled receptor F2; G protein-coupled receptor 111; GPR111; G-protein coupled receptor 111; G-protein coupled receptor PGR20; hGPCR35; PGR20; probable G-protein coupled receptor 111 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZF7 | |
| ADGRF2 | |
| Recombinant protein corresponding to Human GPR111. Recombinant protein control fragment (Product #RP-94921). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction