missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR111 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24965-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| GPR111 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF2 | |
| G protein coupled receptor 111, GPR111, hGPCR35, PGR20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 222611 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IZF7 | |
| ADGRF2 | |
| GPR111 Fusion Protein Ag21705 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction