missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR115 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533610
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Bovine, Horse (100%) Elephant, Dog, Rabbit (94%).
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR115 is a member of this family (subfamily Orphan-B). ESTs have been isolated from breast, heart, and placenta libraries.
Specifications
| GPR115 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF4 | |
| 4632435A09Rik; ADGRF4; Adhesion G protein-coupled receptor F4; AV239010; FLJ38076; G protein-coupled receptor 115; GPR115; G-protein coupled receptor 115; G-protein coupled receptor PGR18; PGR18; probable G-protein coupled receptor 115; RGD1565216; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100068555, 221393, 517950, 78249 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8IZF3, Q9D2L6 | |
| ADGRF4 | |
| Synthetic 16 amino acid peptide from C-terminus of human GPR115. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Monkey, Bovine, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction