missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR116 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1519
This item is not returnable.
View return policy
Description
GPR116 Polyclonal antibody specifically detects GPR116 in Human, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GPR116 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| FLJ90640, G protein-coupled receptor 116, Ig-Hepta homolog, KIAA0758DKFZp564O1923, KPG_001, probable G-protein coupled receptor 116 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human GPR116. | |
| 0.05 mL | |
| GPCR | |
| 221395 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 6 μg/mL | |
| Q8IZF2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Primate, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction