missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR119 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16648534
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16648534 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16648534 Supplier Invitrogen™ Supplier No. PA5102060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR119.

GPCR GPR119 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR119
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR119
Gene Accession No. Q7TQP3, Q8TDV5
Gene Alias G protein-coupled receptor 119; glucose-dependent insulinotropic receptor; GPCR2; Gpr119; G-protein coupled receptor 119; G-protein coupled receptor 2; HGPCR2; RP1-20I3.4
Gene Symbols GPR119
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR119(Accession Q8TDV5), corresponding to amino acid residues S193-I243.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139760, 236781
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.