missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR12 Antibody (584940), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6738J
This item is not returnable.
View return policy
Description
GPR12 Monoclonal antibody specifically detects GPR12 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| GPR12 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| 584940 | |
| Flow Cytometry, Immunohistochemistry | |
| FLJ18149, FLJ97704, G protein-coupled receptor 12, GPCR12, GPCR21, G-protein coupled receptor 12, MGC138349, MGC138351 | |
| NS0 mouse myeloma cell line transfected with human GPR12, Met1-Val334, Accession # P47775 | |
| 0.1 mL | |
| GPCR | |
| 2835 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction