missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR128 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17157713
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17157713 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17157713 Supplier Invitrogen™ Supplier No. PA5109783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LGIWRIVIRI QRGKSTSSSS TPTEFCRNGG TWENGRCICT EEWKGLRCTI ANFCENSTYM GFTFARIPVG RYGPSLQTCG KDTPNAGNPM AVRLCSLSLY GEIELQKVT.

GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR128
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ADGRG7
Gene Accession No. Q8BM96, Q96K78
Gene Alias 9130020O16Rik; Adgrg7; adhesion G protein-coupled receptor G7; adhesion G-protein coupled receptor G7; FLJ14454; FLJ16382; FLJ29035; G protein-coupled receptor 128; GPR128; G-protein coupled receptor 128; MGC142011; MGC163260; probable G-protein coupled receptor 128
Gene Symbols ADGRG7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-135 of human GPR128 (NP_116176.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239853, 304014, 84873
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.