missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR133 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16627424
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16627424 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16627424 Supplier Invitrogen™ Supplier No. PA5100940

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR133.

GPRs are characterized by the presence of 7 transmembrane domains. GPR133 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR133
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRD1
Gene Accession No. Q6QNK2
Gene Alias Adgrd1; adhesion G protein-coupled receptor D1; adhesion G-protein coupled receptor D1; DKFZp434B1272; E230012M21Rik; FLJ16770; G protein-coupled receptor 133; GPR133; G-protein coupled receptor 133; G-protein coupled receptor PGR25; MGC138512; MGC138514; Pgr25; probable G-protein coupled receptor 133
Gene Symbols ADGRD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRD1(Accession Q6QNK2), corresponding to amino acid residues E466-S516.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283383
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.