missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR135 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560979
This item is not returnable.
View return policy
Description
Immunogen sequence: PNISMLLGRN REEGYRTRNV DAFLPSQGPG LQARSRSRLR NRYANRLGAC NRMSSSNPAS GVAGDVAMWA RKNPVVLFCR EGPPEPVTAV TKQP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
Orphan receptor. Has spontaneous activity for beta-arrestin recruitment. Shows a reciprocal regulatory interaction with the melatonin receptor MTNR1B most likely through receptor heteromerization.
Specifications
| GPR135 | |
| Polyclonal | |
| Unconjugated | |
| GPR135 | |
| G protein-coupled receptor 135; GPR135; G-protein coupled receptor 135; HUMNPIIY20; PAFR; probable G-protein coupled receptor 135 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64582 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZ08 | |
| GPR135 | |
| Recombinant protein corresponding to Human GPR135. Recombinant protein control fragment (Product #RP-89584). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction