missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR137 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584570
This item is not returnable.
View return policy
Description
Immunogen sequence: LLAQDPGGRG YPLLWPAPCC SCHSELVP Highest antigen sequence indentity to the following orthologs: Mouse - 44%, Rat - 46%.
GPR137 is a protein coding gene.
Specifications
| GPR137 | |
| Polyclonal | |
| Unconjugated | |
| GPR137 | |
| AI428855; C11orf4; G protein-coupled receptor 137; G protein-coupled receptor TM7SF1L1; GPR137; GPR137A; integral membrane protein GPR137; TM7SF1L1; transmembrane 7 superfamily member 1-like 1 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96N19 | |
| GPR137 | |
| Recombinant protein corresponding to Human GPR137. Recombinant protein control fragment (Product #RP-103475). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction