missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR137 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533626
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Elephant, Panda, Horse, Pig (100%) Hamster (81%).
GPR137 is a protein coding gene.
Specifications
| GPR137 | |
| Polyclonal | |
| Unconjugated | |
| GPR137 | |
| AI428855; C11orf4; G protein-coupled receptor 137; G protein-coupled receptor TM7SF1L1; GPR137; GPR137A; integral membrane protein GPR137; TM7SF1L1; transmembrane 7 superfamily member 1-like 1 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100050142, 100524971, 107173, 505015, 56834, 612124, 689984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q17QQ5, Q80ZU9, Q96N19 | |
| GPR137 | |
| Synthetic 16 amino acid peptide from 2nd cytoplasmic domain of human GPR137. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction