missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR139 Polyclonal Antibody

Description
Immunogen sequence: ISKRFRTMAA ATLKAFFKCQ KQPVQFYTNH NFSITSSPWI SPANSHCIKM LVYQYDK Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
Orphan receptor that seems to act through a G(q/11)-mediated pathway.
Specifications
Specifications
| Antigen | GPR139 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GPR139 |
| Gene Accession No. | Q6DWJ6 |
| Gene Alias | G protein-coupled receptor 139; G protein-coupled receptor PGR3; g(q)-coupled orphan receptor GPRg1; Gm495; Gpr139; GPRG1; G-protein coupled receptor 139; G-protein-coupled receptor PGR3; orphan G protein-coupled receptor TRHRL2; PGR3; Probable G-protein coupled receptor 139 |
| Gene Symbols | GPR139 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?