missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR15 Polyclonal Antibody

GPR15 Polyclonal Antibody
Brand: Invitrogen PA533642
This item is not returnable.
View return policy
Beschreibung
GPR15 Polyclonal Antibody for IHC (P)
Spezifikation
| GPR15 | |
| Polyclonal | |
| Unconjugated | |
| GPR15 | |
| 4933439K08Rik; BoB; brother of Bonzo; G protein-coupled receptor 15; GPR15; Gpr-15; G-protein coupled receptor 15; MGC126828; MGC126830 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P49685 | |
| GPR15 | |
| Synthetic 19 amino acid peptide from 2nd extracellular domain of human GPR15. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction