missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR155 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA557547

Product Code. 15755441

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HLIILPFKRR LEFLWNNKDT AENRDSPVSE EIKMTCQQFI HYHRDLCIRN IVKERRCGAK TSAGTFCGCD LVSW Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

GPR155, an integral membrane protein related to G-protein coupled receptors, has been shown to have specific roles in Huntington disease and autism spectrum disorders.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPR155
Polyclonal
Unconjugated
Gpr155
1110017O10Rik; DEP.7; DEPDC3; F730029F15Rik; G protein-coupled receptor 155; GPR155; GPR155 variant 3; GPR155 variant 6; G-protein coupled receptor PGR22; integral membrane protein GPR155; PGR22
Rabbit
Antigen affinity chromatography
RUO
151556
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7Z3F1
Gpr155
Recombinant protein corresponding to Human GPR155. Recombinant protein control fragment (Product #RP-96181).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.