missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR15L Antibody (1020904) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB104161A
This item is not returnable.
View return policy
Description
GPR15L Monoclonal antibody specifically detects GPR15L in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| GPR15L | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 387695 | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA, Flow Cytometry (Intracellular Staining), CyTOF | |
| 1020904 | |
| ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| C10orf99 chromosome 10 open reading frame 99, UNQ1833 | |
| E. coli-derived human C10orf99 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction