missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR160 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575327
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
GPR160, part of the Class A family of G protein-coupled receptors (GPCRs), is prominently expressed in mouse embryonic stem cells and has been associated with various human solid tumors, particularly prostate cancer. The gene's expression contributes to the self-renewal and pluripotency of stem cells by participating in critical signaling pathways, including the JAK/STAT pathway, which is crucial for maintaining the stem cell cycle. High levels of GPR160 expression are observed in prostate cancer, though its role in disease progression is independent of the CARTp-mediated signaling pathway. Investigations into GPR160 have revealed its potential as a biomarker for prostate cancer. Additionally, it regulates mycobacteria entry into macrophages, implicating its functionality in immune responses. The gene's mechanisms of action, especially within cancer contexts, are subjects of ongoing research aimed at understanding its precise contributions to cellular proliferation and disease phenotypes.
Specifications
| GPR160 | |
| Polyclonal | |
| Unconjugated | |
| GPR160 | |
| 1700025D19Rik; G protein-coupled receptor 160; GPCR1; GPCR150; GPR160; G-protein coupled receptor GPCR1; hGPCR1; probable G-protein coupled receptor 160; putative G protein-coupled receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26996, 499588, 71862 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| Q3U3F9, Q66H29, Q9UJ42 | |
| GPR160 | |
| Synthetic peptide corresponding to amino acids 305-338 of Human GPR160. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction