missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR162 Antibody (1040325) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 463.00
Specifications
| Antigen | GPR162 |
|---|---|
| Clone | 1040325 |
| Concentration | 1.0 mg/mL |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687194
|
Novus Biologicals
MAB10844 |
0.1 mg |
€ 463.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673865
|
Novus Biologicals
MAB10844SS |
0.025 mg |
€ 220.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR162 Monoclonal antibody specifically detects GPR162 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| GPR162 | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| A-2, FLJ78837, G protein-coupled receptor 162, Gene-rich cluster gene A protein, GRCAgene rich cluster, A, probable G-protein coupled receptor 162 | |
| Partial synthetic peptide made to an extracellular loop of the human GPR162 protein (between amino acids 1-100) [UniProt Q16538] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1040325 | |
| Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuroscience | |
| PBS | |
| 27239 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title