missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Monoclonal Antibody (OTI6E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811837S
This item is not returnable.
View return policy
Description
GPR17 Monoclonal antibody specifically detects GPR17 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GPR17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30731 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 140595, 14608 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI6E2 | |
| Unconjugated | |
| GPR83 | |
| AI853548; DKFZp686M18273; G protein-coupled receptor 17; Gpr17; gpr17 {ECO:0000312; G-protein coupled receptor 17; P2Y-like receptor; R12; RGD:1589785}; UDP/CysLT receptor; Uracil nucleotide/cysteinyl leukotriene receptor; Y12546; Z94155 | |
| Synthetic peptide | |
| 30 μL | |
| Primary | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction