missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR171 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA584548
This item is not returnable.
View return policy
Description
Immunogen sequence: LSKAFRSKVT ETFASPKETK AQKEKLRCEN NA
GPR171 is a protein coding gene. Orphan receptor.
Specifications
| GPR171 | |
| Polyclonal | |
| Unconjugated | |
| GPR171 | |
| H963 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29909 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14626 | |
| GPR171 | |
| Recombit protein corresponding to Human GPR171 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction