missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR176 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15706891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15706891 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15706891 Supplier Invitrogen™ Supplier No. PA559018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PPLSTVDSVS QVAPAAPVEP ETFPDKYSLQ FGFGPFELPP QWLSETRNSK KRLLPPLGNT PEELIQTKVP KVGRVERKMS RNNKV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 89%.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR176 (G protein-coupled receptor 176), also known as HB-954, GPR or Gm1012, is a 515 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 1 family. Expressed in brain and spleen, with trace expression in kidney, GPR176 functions as an orphan receptor that is thought to play a role in signaling events throughout the cell. Containing four N-glycosylation sites, seven transmembrane domains and a large C-terminal cytosolic domain, GPR176 is encoded by a gene mapping to human chromosome 15q14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR176
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR176
Gene Accession No. Q14439
Gene Alias Agr9; G protein-coupled receptor 176; Gm1012; Gpr; GPR176; G-protein coupled receptor 176; G-protein coupled receptor AGR9; HB954; HB-954; probable G-protein coupled receptor 176; putative G protein coupled receptor; RBU-15
Gene Symbols GPR176
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR176. Recombinant protein control fragment (Product #RP-102880).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.