Learn More
GPR177/WLS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35821-20ul
 View more versions of this product
Product Code. 30230888
  • € 190.00 / 20µL
Description
GPR177/WLS Polyclonal antibody specifically detects GPR177/WLS in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR177/WLS
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
C1orf139, DKFZp686I0788, EVIchromosome 1 open reading frame 139, FLJ23091, G protein-coupled receptor 177, GPR177wls, Integral membrane protein GPR177, MGC131760, MGC14878, MRP, Protein evenness interrupted homolog, protein wntless homolog, Putative NF-kappa-B-activating protein 373, putative NFkB activating protein 373, wntless homolog (Drosophila)
Recombinant fusion protein containing a sequence corresponding to amino acids 43-232 of human GPR177/WLS (NP_001002292.3).,, Sequence:, YMSVKCVDARKNHHKTKWFVPWGPNHCDKIRDIEEAIPREIEANDIVFSVHIPLPHMEMSPWFQFMLFILQLDIAFKLNNQIRENAEVSMDVSLAYRDDAFAEWTEMAHERVPRKLKCTFTSPKTPEHEGRYYECDVLPFMEIGSVAHKFYLLNIRLPVNEKKKINVGIGEIKDIRLVGIHQNGGFTKVW
20 μL
Signal Transduction
79971
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol