missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR18 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30475.100uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against partial recombinant human GPR18.
- High specificity
- High reactivity
Specifications
| GPR18 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant human GPR18. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| GPR18 | |
| Recombinant protein corresponding to human GPR18. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| Q14330 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2841 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction