Learn More
GPR19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-87572
 View more versions of this product
Product Code. 18756903
  • € 572.00 / 0.10mL
Description
GPR19 Polyclonal specifically detects GPR19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
RUO
IgG
Rabbit
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.