Learn More
Invitrogen™ GPR21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532817
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%); Rat (88%); Mouse, Pig, Bovine, Bat (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids b chain (active). HuFSH induces maturation of Graafian follicles of the ovary, promotes development of germinal cells in males and activates cytosolic tyrosine kinase. The HuFSH is purified by proprietary chromatographic techniques.
Specifications
| GPR21 | |
| Polyclonal | |
| Unconjugated | |
| GPR21 | |
| C230004C13Rik; G protein-coupled receptor 21; G protein-coupled receptor GPR21; Gpr21; Probable G-protein coupled receptor 21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q99679 | |
| GPR21 | |
| Synthetic 16 amino acid peptide from C-terminus of human GPR21. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.