missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR22 Antibody (455108), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4500J
This item is not returnable.
View return policy
Description
GPR22 Monoclonal antibody specifically detects GPR22 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| GPR22 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 455108 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22 | |
| HEK293 human embryonic kidney cell line transfected with human GPR22, Met1-Asp433, Accession # Q99680 | |
| 0.1 mL | |
| GPCR | |
| 2845 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction