missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR34 Antibody (419859), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4617M
This item is not returnable.
View return policy
Description
GPR34 Monoclonal antibody specifically detects GPR34 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| GPR34 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 419859 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| G protein-coupled receptor 34, probable G-protein coupled receptor 34 | |
| NS0 mouse myeloma cell line transfected with human GPR34, Accession # Q9UPC5 | |
| 0.1 mL | |
| GPCR | |
| 2857 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction