missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR39 Antibody (189702) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3348B
This item is not returnable.
View return policy
Description
GPR39 Monoclonal antibody specifically detects GPR39 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| GPR39 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 189702 | |
| Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541 | |
| NS0 mouse myeloma cell line transfected with human GPR39, Met1-Val453, Accession # BAG37409 | |
| 0.1 mL | |
| GPCR, Lipid and Metabolism | |
| 2863 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction