missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533709
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon (95%) Marmoset (85%).
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. G protein-coupled receptor 39 is an orphan receptor in the Ghrelin family pathway. Various studies have shown that GPR39 plays important roles in hypertension, adipogenesis and that overexpression of GPR39 contributes to malignant development of human esophageal squamous cell carcinoma.
Specifications
| GPR39 | |
| Polyclonal | |
| Unconjugated | |
| GPR39 | |
| 4933415E13Rik; G protein-coupled receptor 39; GPR39; G-protein coupled receptor 39 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2863 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| PBS with 0.1% sodium azide | |
| O43194 | |
| GPR39 | |
| Synthetic 20 amino acid peptide from C-terminus of human GPR39. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction