missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR50 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533732
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Sheep, Dog (100%) Gibbon, Mouse, Rat, Panda, Bovine, Rabbit (94%) Bat (88%).
Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.
Specifications
| GPR50 | |
| Polyclonal | |
| Unconjugated | |
| GPR50 | |
| G protein-coupled receptor 50; Gpr50; G-protein-coupled receptor 50; H9; Mel1c; melatonin-related receptor; MTNRL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 443023, 492213, 9248 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q13585, Q28558 | |
| GPR50 | |
| Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR50. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Sheep | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction