missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR52 Antibody (582671) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10357JF669
This item is not returnable.
View return policy
Description
GPR52 Monoclonal antibody specifically detects GPR52 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| GPR52 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| G protein-coupled receptor 52, MGC111751, probable G-protein coupled receptor 52 | |
| NS0 mouse myeloma cell line transfected with human GPR52, Met1-Ile361, Accession # Q9Y2T5 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 582671 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 9293 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction