missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR55 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-55498
This item is not returnable.
View return policy
Description
GPR55 Polyclonal antibody specifically detects GPR55 in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| GPR55 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| G protein-coupled receptor 55, G-protein coupled receptor 55 | |
| Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human GPR55. | |
| 0.05 mg | |
| GPCR | |
| 9290 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin 5 μg/mL, Immunohistochemistry-Frozen | |
| Q9Y2T6 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction