missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR55 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ OPA115577
This item is not returnable.
View return policy
Description
By immunohistochemistry, OPA1-15577 detects GPR55 in formalin fixed, paraffin embedded human tissues.
GPR55 has been reported to be expressed in the brain, where mRNA transcripts were detected in the caudate nucleus and putamen, but not in five other brain regions (Sawzdargo et al., 1999). ESTs have been isolated from liver/spleen libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This 'turn-off' activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Twoblockbuster drugs which have been developed as GPCR-targetedpharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
| GPR55 | |
| Polyclonal | |
| Unconjugated | |
| GPR55 | |
| G protein-coupled receptor 55; Gm218; GPCR55; GPR55; G-protein coupled receptor 55; Lpir1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9290 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y2T6 | |
| GPR55 | |
| Synthetic 17 amino acid peptide from 3rd extracellular domain of human GPR55. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction