missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR56 Antibody (776123), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4636Z
This item is not returnable.
View return policy
Description
GPR56 Monoclonal antibody specifically detects GPR56 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| GPR56 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 776123 | |
| Flow Cytometry, CyTOF-ready | |
| BFPP, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4G-protein coupled receptor 56, TM7XN1DKFZp781L1398 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val342, Accession # Q9Y653 | |
| 0.1 mL | |
| GPCR, Growth and Development, Neuronal Cell Markers | |
| 9289 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction