missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR56 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1214
This item is not returnable.
View return policy
Description
GPR56 Polyclonal antibody specifically detects GPR56 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GPR56 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| BFPP, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4G-protein coupled receptor 56, TM7XN1DKFZp781L1398 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR56. | |
| 0.05 mL | |
| GPCR | |
| 9289 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 4 - 32 μg/mL | |
| Q9Y653 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction