missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR68 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118514
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OGR1/GPR68.
GPR68 (also known as OGR1 - ovarian cancer G-protein coupled receptor 1) is a proton-sensing receptor involved in pH homeostasis. It represents an osteoblastic pH sensor regulating the cell-mediated response to acidosis in bone. GPR68 mediates its action by association with G proteins that stimulate inositol phosphate (IP) production or CA2+ mobilization. The receptor is almost silent at pH 7.8 but fully activated at pH 6.8. Also found to serve as a tumor suppressor in prostate cancer. Mutations to the gene result in amelogenesis imperfecta, hypomaturation type 2A6.
Specifications
| GPR68 | |
| Polyclonal | |
| Unconjugated | |
| GPR68 | |
| BB131428; Brgr1; Brgrb; G protein-coupled receptor 68; GPR12A; Gpr68; G-protein coupled receptor 68; Ogr1; OGR-1; ovarian cancer G protein-coupled receptor, 1; ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 238377, 8111 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15743, Q8BFQ3 | |
| GPR68 | |
| A synthesized peptide derived from human GPR68(Accession Q15743), corresponding to amino acid residues F194-Y244. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction