missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR68 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533758
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Bat, Horse, Rabbit, Pig (100%).
GPR68 (also known as OGR1 - ovarian cancer G-protein coupled receptor 1) is a proton-sensing receptor involved in pH homeostasis. It represents an osteoblastic pH sensor regulating the cell-mediated response to acidosis in bone. GPR68 mediates its action by association with G proteins that stimulate inositol phosphate (IP) production or CA2+ mobilization. The receptor is almost silent at pH 7.8 but fully activated at pH 6.8. Also found to serve as a tumor suppressor in prostate cancer. Mutations to the gene result in amelogenesis imperfecta, hypomaturation type 2A6.
Specifications
| GPR68 | |
| Polyclonal | |
| Unconjugated | |
| GPR68 | |
| BB131428; Brgr1; Brgrb; G protein-coupled receptor 68; GPR12A; Gpr68; G-protein coupled receptor 68; Ogr1; OGR-1; ovarian cancer G protein-coupled receptor, 1; ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100053071, 100153615, 100752950, 103351698, 238377, 314386, 8111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15743, Q8BFQ3 | |
| GPR68 | |
| Synthetic 18 amino acid peptide from 2nd cytoplasmic domain of human GPR68. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Monkey, Hamster, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction