missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR75 Polyclonal Antibody

GPR75 Polyclonal Antibody
Brand: Invitrogen PA533762
This item is not returnable.
View return policy
Description
GPR75 Polyclonal Antibody for IHC (P)
Specifications
| GPR75 | |
| Polyclonal | |
| Unconjugated | |
| GPR75 | |
| G protein-coupled receptor 75; GPR75; GPRchr2; GPR-chr2; Probable G-protein coupled receptor 75; WI31133; WI-31133 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100066961, 100517153, 10936, 539470 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O95800 | |
| GPR75 | |
| Synthetic 16 amino acid peptide from 2nd cytoplasmic domain of human GPR75. | |
| 50 μg | |
| Primary | |
| Human, Bovine, Pig, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction