missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR75 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17972-1-AP-20UL
This item is not returnable.
View return policy
Description
GPR75 is an Orphan-A GPCR with an unknown ligand. G protein-coupled receptor GPR75 has been reported to be expressed in brain and eye. ESTs have been isolated from human brain, eye, nerve, skin, and stomach libraries.Specifications
| GPR75 | |
| Polyclonal | |
| Unconjugated | |
| GPR75 | |
| G protein coupled receptor 75, GPR chr2, GPR75, WI 31133 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10936, 237716 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95800, Q6X632 | |
| Gpr75 | |
| GPR75 Fusion Protein Ag12053 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction