missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR81 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-02369
This item is not returnable.
View return policy
Description
GPR81 Polyclonal antibody specifically detects GPR81 in Human, Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GPR81 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| FKSG80, G protein-coupled receptor 104, G protein-coupled receptor 81, GPR104, G-protein coupled receptor 104, G-protein coupled receptor 81, HCA1, HCAR1, LACR1, lactate receptor 1, TA-GPCR, T-cell activation G protein-coupled receptor | |
| Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GPR81. | |
| 0.05 mg | |
| GPCR, Immunology | |
| 27198 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| ELISA 1:100 - 1:2000, Immunohistochemistry 20 μg/mL, Immunohistochemistry-Paraffin 20 μg/mL | |
| Q9BXC0 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction