missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR83 Monoclonal Antibody (OTI6C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811749S
This item is not returnable.
View return policy
Description
GPR83 Monoclonal antibody specifically detects GPR83 in Human samples. It is validated for Western BlotSpecifications
| GPR83 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13304 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2840 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI6C6 | |
| Unconjugated | |
| GPR17 | |
| AW045253; G protein-coupled receptor 72; G protein-coupled receptor 83; Gir; glucocorticoid induced receptor; glucocorticoid-induced receptor; GPR72; Gpr83; G-protein coupled receptor 72; Hcepto9; Jp05; KIAA1540; Probable G-protein coupled receptor 83; RP105; RP39; RP82 | |
| Synthetic peptide (the amino acid sequence is considered to be commercially sensitive) within Human GPR17 C-terminal. The exact sequence is proprietary. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction